Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium(bromomethyl)trifluoroborate ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

048489-1G 048489-5G
101884-060EA 16.92 USD InStock
101884-060 101884-062
Potassium(bromomethyl)trifluoroborate ≥95% for synthesis
Potassium(bromomethyl)trifluoroborate
Hazard code(s): Irritant


Formula: CH₂BBrF₃K
MW: 200.83 g/mol
Melting Pt: 225…230 °C
MDL Number: MFCD09265154
CAS Number: 888711-44-2
Order Now
Specification Test Results
Melting point 225-230°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty