Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorobenzamidine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

056392-500MG 056392-1G
101910-322EA 156.92 USD InStock
101910-322 101910-324
3,5-Difluorobenzamidine hydrochloride ≥95%
3,5-Difluorobenzamidine hydrochloride
Hazard code(s): Irritant


Formula: C₇H₇ClF₂N₂
MDL Number: MFCD04114449
CAS Number: 144797-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty