Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Tribromophenyl isothiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

056623-1G 056623-5G
101911-074EA 69.23 USD InStock
101911-074 101911-076
2,4,6-Tribromophenyl isothiocyanate ≥98%
2,4,6-Tribromophenyl isothiocyanate
Hazard code(s): Irritant-HARMFUL


Formula: C₇H₂Br₃NS
Boiling Pt: 142 °C (0.05 -rr)
Melting Pt: 107 - 108 °C
MDL Number: MFCD00041039
CAS Number: 22134-11-8
UN: 2811
ADR: 6.1,I
Order Now
Specification Test Results
Melting point 106-110°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty