Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Adipohydrazide for synthesis

Supplier: MATRIX SCIENTIFIC MS

056879-10G 056879-50G
101911-576EA 15.38 USD InStock
101911-576 101911-578
Adipohydrazide for synthesis
Adipohydrazide
Hazard code(s): Irritant


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 446 °C (1013 hPa)
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00007614
CAS Number: 1071-93-8
EINECS: 213-999-5
Order Now
Specification Test Results
Melting point 180-182°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty