Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromobenzhydrazide

Supplier: MATRIX SCIENTIFIC MS

056900-10G 056900-50G
101911-652EA 156.92 USD InStock
101911-652 101911-654
3-Bromobenzhydrazide
3-Bromobenzhydrazide
Hazard code(s): Irritant


Formula: C₇H₇BrN₂O
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 151 °C
MDL Number: MFCD00007600
CAS Number: 39115-96-3
EINECS: 254-298-4
Order Now
Specification Test Results
Melting point 155-156°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty