Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-methoxybenzoic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

056958-25G 056958-100G
101911-874EA 223.08 USD InStock
101911-874 101911-876
3-Chloro-4-methoxybenzoic acid ≥98% for synthesis
3-Chloro-4-methoxybenzoic acid
Hazard code(s): Irritant


Formula: C₈H₇ClO₃
MW: 186.59
Melting Pt: 213…215 °C
MDL Number: MFCD00016512
CAS Number: 37908-96-6
EINECS: 253-708-9
Order Now
Specification Test Results
Melting point 213-215°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty