Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-o-xylene ≥98%

Supplier: MATRIX SCIENTIFIC MS

056987-25G 056987-100G
101911-972EA 252.31 USD InStock
101911-972 101911-974
4-Chloro-o-xylene ≥98%
4-Chloro-o-xylene
Hazard code(s): Irritant


Formula: C₈H₉Cl
Boiling Pt: 194 - 195 °C (1013 hPa)
Melting Pt: -6 °C
Density: 1.0682 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000596
CAS Number: 615-60-1
EINECS: 210-438-6
Order Now
Specification Test Results
Boiling point 221-223°C
Density 1.05

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty