Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-4-hydroxybenzoic acid for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

056994-25G 056994-100G
101911-990EA 95.38 USD InStock
101911-990 101911-992
3,5-Dibromo-4-hydroxybenzoic acid for synthesis
3,5-Dibromo-4-hydroxybenzoic acid
Hazard code(s): Irritant


Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Boiling Pt: 365
Melting Pt: 271…273 °C
MDL Number: MFCD00002548
CAS Number: 3337-62-0
EINECS: 222-075-0
Order Now
Specification Test Results
Melting point 271-273°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty