Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Dichlorophenyl)-2-thiourea for synthesis

Supplier: MATRIX SCIENTIFIC MS
Danger

057013-10G 057013-50G
101912-044EA 109.23 USD InStock
101912-044 101912-046
1-(3,4-Dichlorophenyl)-2-thiourea for synthesis
1-(3,4-Dichlorophenyl)-2-thiourea
Hazard code(s): Irritant


Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 197…199 °C
MDL Number: MFCD00004935
CAS Number: 19250-09-0
EINECS: 242-919-1
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 197-199°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty