Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',5'-Dimethoxyacetophenone ≥99%

Supplier: MATRIX SCIENTIFIC MS

057025-25G 057025-100G
101912-088EA 198.46 USD InStock
101912-088 101912-090
2',5'-Dimethoxyacetophenone ≥99%
2',5'-Dimethoxyacetophenone
Hazard code(s): Irritant


Formula: C₁₀H₁₂O₃
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 18 - 20 °C
Density: 1.139 g/cm³ (20 °C)
MDL Number: MFCD00008728
CAS Number: 1201-38-3
EINECS: 214-858-0
Order Now
Specification Test Results
Boiling point 155°C /10mm
Melting point 18-20°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty