Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorophenol ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

058195-25G 058195-100G
101914-178EA 41.54 USD InStock
101914-178 101914-180
3,5-Dichlorophenol ≥98%
3,5-Dichlorophenol
Hazard code(s): Toxic


Formula: C₆H₄Cl₂O
Boiling Pt: 233 °C
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002259
CAS Number: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty