Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Thiazolidinedione

Supplier: MATRIX SCIENTIFIC MS

058274-25G 058274-100G
101914-488EA 21.54 USD InStock
101914-488 101914-490
2,4-Thiazolidinedione
2,4-Thiazolidinedione
Hazard code(s): Irritant


Formula: C₃H₃NO₂S
Melting Pt: 123…126 °C
MDL Number: MFCD00005478
CAS Number: 2295-31-0
EINECS: 218-941-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty