Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyridone for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

058298-100G 058298-250G
101914-534EA 75.38 USD InStock
101914-534 101914-536
4-Pyridone for synthesis
4-Pyridone
Hazard code(s): Irritant


Formula: C₅H₅NO
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 146 - 148 °C
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3
Order Now
Specification Test Results
Melting point 143-145

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty