Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl isopropylmalonate for synthesis

Supplier: MATRIX SCIENTIFIC MS

058362-25G 058362-100G
101914-656EA 44.62 USD InStock
101914-656 101914-658
Diethyl isopropylmalonate for synthesis
Diethyl isopropylmalonate
Hazard code(s): Irritant


Formula: C₁₀H₁₈O₄
Boiling Pt: 102…104 °C (12mm)
Density: 0.981 g/cm³
MDL Number: MFCD00040491
CAS Number: 759-36-4
EINECS: 212-068-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty