Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromovanillin

Supplier: MATRIX SCIENTIFIC MS
Warning

058396-25G 058396-100G
101914-724EA 52.31 USD InStock
101914-724 101914-726
5-Bromovanillin
5-Bromovanillin


Formula: C₈H₇BrO₃
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 166…167 °C
MDL Number: MFCD00006940
CAS Number: 2973-76-4
EINECS: 221-016-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty