Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-4-(trifluoromethyl)aniline ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

066904-5G 066904-25G
101932-714EA 21.54 USD InStock
101932-714 101932-716
2-Iodo-4-(trifluoromethyl)aniline ≥98% for synthesis
2-Iodo-4-(trifluoromethyl)aniline
Hazard code(s): Irritant


Formula: C₇H₅F₃IN
MW: 287.02
MDL Number: MFCD02683097
CAS Number: 163444-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty