Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chloroanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

066950-5G 066950-25G
101932-846EA 23.08 USD InStock
101932-846 101932-848
5-Bromo-2-chloroanisole ≥98%
5-Bromo-2-chloroanisole
Hazard code(s): Irritant


Formula: C₇H₆BrClO
Boiling Pt: 240…245 °C
Melting Pt: 27…28 °C
Density: 1.631 g/cm³
MDL Number: MFCD00078659
CAS Number: 16817-43-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty