Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromothiophene ≥92% for synthesis

Supplier: MATRIX SCIENTIFIC MS

067224-5G 067224-25G
101933-576EA 66.15 USD InStock
101933-576 101933-578
2,4-Dibromothiophene ≥92% for synthesis
2,4-Dibromothiophene
Hazard code(s): Toxic


Formula: C₄H₂Br₂S
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: -30 °C
Density: 2.14 g/cm³ (20 °C)
MDL Number: MFCD00043889
CAS Number: 3140-92-9
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty