Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2-carboxylic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

069602-25G 069602-100G
101941-026EA 23.08 USD InStock
101941-026 101941-028
Pyrazine-2-carboxylic acid ≥98% for synthesis
Pyrazine-2-carboxylic acid
Hazard code(s): Irritant


Formula: C₅H₄N₂O₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 222…225 °C (dec.)
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1
Merck Index: 12,08143
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty