Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Ethylthio)phenylboronic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

069728-1G 069728-5G 069728-25G
101941-644EA 524.62 USD InStock
101941-644 101941-646 101941-648
3-(Ethylthio)phenylboronic acid ≥98% for synthesis
3-(Ethylthio)phenylboronic acid
Hazard code(s): Irritant


Formula: C₈H₁₁BO₂S
MDL Number: MFCD03092930
CAS Number: 870718-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty