Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carnegine

Supplier: MATRIX SCIENTIFIC MS

001044-100MG 001044-250MG
101780-882EA 153.85 USD InStock
101780-882 101780-884
Carnegine
Carnegine
Hazard: Irritant


Formula: C₁₃H₁₉NO₂
MW: 221.3 g/mol
Melting Pt: 207 °C
CAS Number: 5852-92-6
Order Now
Specification Test Results
Melting point 207°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty