Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(bromomethyl)-3-methoxybenzoate

Supplier: MATRIX SCIENTIFIC MS

001088-5G 001088-25G
101780-918EA 52.31 USD InStock
101780-918 101780-920
Methyl-4-(bromomethyl)-3-methoxybenzoate
Methyl 4-(bromomethyl)-3-methoxybenzoate
Hazard: Toxic


Formula: C₁₀H₁₁BrO₃
MW: 259.0965 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD00270115
CAS Number: 70264-94-7
Order Now
Specification Test Results
Melting point 92-94°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty