Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-methylphenylacetic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

001462-1G 001462-5G
101781-720EA 27.69 USD InStock
101781-720 101781-722
4-Methoxy-3-methylphenylacetic acid ≥98%
4-Methoxy-3-methylphenylacetic acid
Hazard: Irritant


Formula: C₁₀H₁₂O₃
Melting Pt: 128°C
MDL Number: MFCD00041034
CAS Number: 4513-73-9
Order Now
Specification Test Results
Melting point 93-95°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty