Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline

Supplier: MATRIX SCIENTIFIC MS

001517-250MG 001517-1G
101781-868EA 87.69 USD InStock
101781-868 101781-870
4-Hydroxy-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
4-Hydroxy-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
Hazard: Irritant


Formula: C₁₁H₅F₆NO₂
Melting Pt: 229…232 °C
MDL Number: MFCD00276584
CAS Number: 125647-79-2
Order Now
Specification Test Results
Melting point 229-232°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty