Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-7-methyl-1,2,4-benzotriazine-1-oxide ≥97%

Supplier: MATRIX SCIENTIFIC MS

001803-1G 001803-5G
101782-634EA 86.15 USD InStock
101782-634 101782-636
3-Amino-7-methyl-1,2,4-benzotriazine-1-oxide ≥97%
3-Amino-7-methyl-1,2,4-benzotriazine-1-oxide
Hazard: Irritant


Formula: C₈H₈N₄O
Melting Pt: 275 °C
MDL Number: MFCD00024042
CAS Number: 27281-74-9
Order Now
Specification Test Results
Melting point 275°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty