Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(2,4-difluorophenyl)thiazole-4-carboxylate

Supplier: MATRIX SCIENTIFIC MS

001944-250MG 001944-1G
101782-904EA 52.31 USD InStock
101782-904 101782-906
Ethyl-2-(2,4-difluorophenyl)thiazole-4-carboxylate
Ethyl-2-(2,4-difluorophenyl)thiazole-4-carboxylate
Hazard: Irritant


Formula: C₁₂H₉F₂NO₂S
Melting Pt: 50…52 °C
MDL Number: MFCD00277200
CAS Number: 175276-93-4
Order Now
Specification Test Results
Melting point 50-52°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty