Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

002032-5G 002032-25G
101783-078EA 29.23 USD InStock
101783-078 101783-080
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid ≥97%
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Hazard: Irritant


Formula: C₉H₆F₄O₃
Melting Pt: 176…177 °C
MDL Number: MFCD00155926
CAS Number: 10009-25-3
Order Now
Specification Test Results
Melting point 176-177°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty