Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid

Supplier: MATRIX SCIENTIFIC MS

002043-250MG 002043-1G
101783-096EA 23.08 USD InStock
101783-096 101783-098
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
Hazard: Irritant


Formula: C₁₂H₈F₃NO₂S
MW: 287.26 g/mol
Melting Pt: 238…241 °C
MDL Number: MFCD00068105
CAS Number: 144059-86-9
Order Now
Specification Test Results
Melting point 238-241°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty