Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-methylphenylhydrazine hydrochloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

002180-1G 002180-5G
101783-404EA 92.31 USD InStock
101783-404 101783-406
5-Fluoro-2-methylphenylhydrazine hydrochloride ≥97%
5-Fluoro-2-methylphenylhydrazine hydrochloride
Hazard: Irritant


Formula: C₇H₁₀ClFN₂
Melting Pt: 196…197 °C
MDL Number: MFCD00053032
CAS Number: 325-50-8
Order Now
Specification Test Results
Melting point 196-197°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty