Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

002612-1G 002612-5G
101784-284EA 269.23 USD InStock
101784-284 101784-286
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥97%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride
Hazard: Irritant


Formula: C₇H₄F₅NO
Melting Pt: 227 °C
MDL Number: MFCD00012953
CAS Number: 57981-02-9
Order Now
Specification Test Results
Melting point 227°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty