Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid

Supplier: MATRIX SCIENTIFIC MS

002646-1G 002646-5G
101784-384EA 29.23 USD InStock
101784-384 101784-386
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Hazard: Irritant


Formula: C₁₁H₇F₃N₂O₂
Melting Pt: 136…139 °C
MDL Number: MFCD00173854
CAS Number: 98534-81-7
Order Now
Specification Test Results
Melting point 136-139°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty