Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-fluorophenol ≥98%

Supplier: MATRIX SCIENTIFIC MS

041030-1G 041030-5G
101850-652EA 183.08 USD InStock
101850-652 101850-654
2-Amino-3-fluorophenol ≥98%
2-Amino-3-fluorophenol
Hazard: Irritant/Light Sensitive/Refrigerate


Formula: C₆H₆FNO
MW: 127.1163 g/mol
Melting Pt: 126…127 °C
MDL Number: MFCD07368754
CAS Number: 53981-23-0
Order Now
Specification Test Results
Melting point 126-127°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty