Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-({2-Oxo-2-[4-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044437-500MG 044437-1G 044437-5G
101865-838EA 181.54 USD InStock
101865-838 101865-840 101865-842
2-({2-Oxo-2-[4-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid ≥95%
2-({2-Oxo-2-[4-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid
Hazard: Irritant


Formula: C₁₁H₁₀F₃NO₃S
MW: 293.27 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00202307
CAS Number: 338421-16-2
Order Now
Specification Test Results
Melting point 121-123°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty