Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate 95

Supplier: MATRIX SCIENTIFIC MS

044442-500MG 044442-1G 044442-5G
101865-868EA 747.69 USD InStock
101865-868 101865-870 101865-872
Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate 95
Methyl-1-(4-fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylate
Hazard: Irritant


Formula: C₁₃H₁₄FNO₄
MW: 267.26 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD00202776
CAS Number: 318247-24-4
Order Now
Specification Test Results
Melting point 150-152°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty