Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Acetylamino)isonicotinic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044484-500MG 044484-1G 044484-5G
101866-110EA 538.46 USD InStock
101866-110 101866-112 101866-114
3-(Acetylamino)isonicotinic acid ≥95%
3-(Acetylamino)isonicotinic acid
Hazard: Irritant


Formula: C₈H₈N₂O₃
Melting Pt: 281…283 °C
MDL Number: MFCD00598516
CAS Number: 215103-12-1
Order Now
Specification Test Results
Melting point 281-283°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty