Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1-(4-chlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

044516-500MG 044516-1G 044516-5G
101866-284EA 375.38 USD InStock
101866-284 101866-286 101866-288
Methyl-1-(4-chlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Methyl-1-(4-chlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate
Hazard: Irritant


Formula: C₁₅H₁₆ClNO₃
Melting Pt: 84…86 °C
MDL Number: MFCD00975047
CAS Number: 338748-81-5
Order Now
Specification Test Results
Melting point 84-86°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty