Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(5-Nitropyridin-2-yl)benzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044657-500MG 044657-1G 044657-5G
101866-950EA 747.69 USD InStock
101866-950 101866-952 101866-954
4-(5-Nitropyridin-2-yl)benzoic acid ≥95%
4-(5-Nitropyridin-2-yl)benzoic acid
Hazard: Irritant


Formula: C₁₂H₈N₂O₄
Melting Pt: 307…309 °C
CAS Number: 223127-49-9
Order Now
Specification Test Results
Melting point 307-309°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty