Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044673-500MG 044673-1G 044673-5G
101867-028EA 375.38 USD InStock
101867-028 101867-030 101867-032
1-(2-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(2-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant


Formula: C₁₂H₁₁BrN₂O₂
Melting Pt: 235…237 °C
MDL Number: MFCD06200972
CAS Number: 959578-21-3
Order Now
Specification Test Results
Melting point 235-237°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty