Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(5-Amino-1H-1,2,4-triazol-3-yl)sulfanyl]acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

044726-500MG 044726-1G 044726-5G
101867-288EA 200 USD InStock
101867-288 101867-290 101867-292
2-[(5-Amino-1H-1,2,4-triazol-3-yl)sulfanyl]acetonitrile ≥95%
2-[(5-Amino-1H-1,2,4-triazol-3-yl)sulfanyl]acetonitrile
Hazard: Irritant


Formula: C₄H₅N₅S
MW: 155.18 g/mol
Melting Pt: 130…133 °C
MDL Number: MFCD00138376
CAS Number: 338751-47-6
Order Now
Specification Test Results
Melting point 130-133°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty