Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044748-500MG 044748-1G 044748-5G
101867-412EA 132.31 USD InStock
101867-412 101867-414 101867-416
2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%
2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant


Formula: C₁₄H₁₀F₃NO₃
Melting Pt: 159…161 °C
MDL Number: MFCD00139983
CAS Number: 338754-66-8
Order Now
Specification Test Results
Melting point 159-161°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty