Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(5-phenyl-2H-1,2,3,4-tetraazol-2-yl)acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

044765-500MG 044765-1G 044765-5G
101867-490EA 375.38 USD InStock
101867-490 101867-492 101867-494
Ethyl-2-(5-phenyl-2H-1,2,3,4-tetraazol-2-yl)acetate ≥95%
Ethyl-2-(5-phenyl-2H-1,2,3,4-tetraazol-2-yl)acetate
Hazard: Irritant


Formula: C₁₁H₁₂N₄O₂
Melting Pt: 83…85 °C
MDL Number: MFCD00268800
CAS Number: 21054-65-9
Order Now
Specification Test Results
Melting point 83-85°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty