Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2H-chromene-3-carboxylic acid methyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

044770-500MG 044770-1G 044770-5G
101867-514EA 603.08 USD InStock
101867-514 101867-516 101867-518
6-Methoxy-2H-chromene-3-carboxylic acid methyl ester ≥95%
6-Methoxy-2H-chromene-3-carboxylic acid methyl ester
Hazard: Irritant


Formula: C₁₂H₁₂O₄
Melting Pt: 82…84 °C
MDL Number: MFCD00202061
CAS Number: 338759-76-5
Order Now
Specification Test Results
Melting point 82-84°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty