Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(5-amino-2,4-dichlorophenoxy)acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

044815-500MG 044815-1G 044815-5G
101867-720EA 556.92 USD InStock
101867-720 101867-722 101867-724
Ethyl-2-(5-amino-2,4-dichlorophenoxy)acetate ≥95%
Ethyl-2-(5-amino-2,4-dichlorophenoxy)acetate
Hazard: Irritant


Formula: C₁₀H₁₁Cl₂NO₃
MW: 264.11 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00974418
CAS Number: 338960-27-3
Order Now
Specification Test Results
Melting point 86-88°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty