Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indole-2-carbothioamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

044843-500MG 044843-1G
101867-846EA 747.69 USD InStock
101867-846 101867-848
1H-Indole-2-carbothioamide ≥97%
1H-Indole-2-carbothioamide
Hazard: Irritant


Formula: C₉H₈N₂S
MW: 176.24 g/mol
Melting Pt: 212…214 °C
MDL Number: MFCD05662471
CAS Number: 63071-71-6
Order Now
Specification Test Results
Melting point 212-214°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty