Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

044845-500MG 044845-1G 044845-5G
101867-856EA 375.38 USD InStock
101867-856 101867-858 101867-860
4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile ≥95%
4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile
Hazard: Irritant


Formula: C₁₄H₈F₃NO
MW: 263.22 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD01871521
CAS Number: 453557-46-5
Order Now
Specification Test Results
Melting point 59-61°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty