Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(2-Carboxyethyl)sulfanyl]nicotinic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044857-500MG 044857-1G 044857-5G
101867-920EA 278.46 USD InStock
101867-920 101867-922 101867-924
2-[(2-Carboxyethyl)sulfanyl]nicotinic acid ≥95%
2-[(2-Carboxyethyl)sulfanyl]nicotinic acid
Hazard: Irritant


Formula: C₉H₉NO₄S
Melting Pt: 203…205 °C
MDL Number: MFCD02250406
CAS Number: 286472-02-4
Order Now
Specification Test Results
Melting point 203-205°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty