Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[(Cyanomethyl)sulfanyl]-5-methyl-4-isothiazolecarbonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

044977-500MG 044977-1G 044977-5G
101868-530EA 278.46 USD InStock
101868-530 101868-532 101868-534
3-[(Cyanomethyl)sulfanyl]-5-methyl-4-isothiazolecarbonitrile ≥95%
3-[(Cyanomethyl)sulfanyl]-5-methyl-4-isothiazolecarbonitrile
Hazard: Irritant


Formula: C₇H₅N₃S₂
Melting Pt: 126…128 °C
MDL Number: MFCD00215371
CAS Number: 338778-08-8
Order Now
Specification Test Results
Melting point 126-128°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty