Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Phenyl-1,3-thiazol-5-yl)-1-ethanone ≥95%

Supplier: MATRIX SCIENTIFIC MS

044982-250MG 044982-1G 044982-5G
101868-560EA 467.69 USD InStock
101868-560 101868-562 101868-564
1-(2-Phenyl-1,3-thiazol-5-yl)-1-ethanone ≥95%
1-(2-Phenyl-1,3-thiazol-5-yl)-1-ethanone
Hazard: Irritant


Formula: C₁₁H₉NOS
MW: 203.26 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00244691
CAS Number: 10045-50-8
Order Now
Specification Test Results
Melting point 151-153°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty