Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(Carboxymethyl)sulfanyl]nicotinic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045092-500MG 045092-1G 045092-5G
101869-138EA 278.46 USD InStock
101869-138 101869-140 101869-142
2-[(Carboxymethyl)sulfanyl]nicotinic acid ≥95%
2-[(Carboxymethyl)sulfanyl]nicotinic acid
Hazard: Irritant


Formula: C₈H₇NO₄S
MW: 213.21 g/mol
Melting Pt: 216…218 °C
MDL Number: MFCD02250402
CAS Number: 325704-15-2
Order Now
Specification Test Results
Melting point 216-218°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty