Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045128-500MG 045128-1G
101869-272EA 747.69 USD InStock
101869-272 101869-274
Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate
Hazard: Irritant


Formula: C₁₆H₁₉N₃O₂
MW: 285.35 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD03425801
CAS Number: 453557-64-7
Order Now
Specification Test Results
Melting point 69-71°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty